2,2',3,3',4,5,6-Heptachlorbiphenyl (PCB Nr. 173)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-173-C
PROOCIL-PCB-173-CEA 862 EUR
PROOCIL-PCB-173-C
2,2',3,3',4,5,6-Heptachlorbiphenyl (PCB Nr. 173)
2,2',3,3',4,5,6-Heptachlorbiphenyl (PCB Nr. 173)
Formel: (C₆H₃Cl₂)C₆Cl₅
Molecular Weight: 395.32 g/mol
CAS-Nummer: 68194-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR