2,2',3,3',4,5,6'-Heptachlorbiphenyl (PCB Nr. 174)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-174-C
PROOCIL-PCB-174-CEA 677 EUR
PROOCIL-PCB-174-C
2,2',3,3',4,5,6'-Heptachlorbiphenyl (PCB Nr. 174)
2,2',3,3',4,5,6'-Heptachlorbiphenyl (PCB Nr. 174)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 38411-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR