2,2',3,3',4,5',6-Heptachlorbiphenyl (PCB Nr. 175)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-175-C
PROOCIL-PCB-175-CEA 713 EUR
PROOCIL-PCB-175-C
2,2',3,3',4,5',6-Heptachlorbiphenyl (PCB Nr. 175)
2,2',3,3',4,5',6-Heptachlorbiphenyl (PCB Nr. 175)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 40186-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR