2,2',3,3',5,5',6-Heptachlorbiphenyl (PCB Nr. 178)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-178-C
PROOCIL-PCB-178-CEA 713 EUR
PROOCIL-PCB-178-C
2,2',3,3',5,5',6-Heptachlorbiphenyl (PCB Nr. 178)
2,2',3,3',5,5',6-Heptachlorbiphenyl (PCB Nr. 178)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 52663-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR