2,2',3,3',5,6,6'-Heptachlorbiphenyl (PCB Nr. 179)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-179-C
PROOCIL-PCB-179-CEA 677 EUR
PROOCIL-PCB-179-C
2,2',3,3',5,6,6'-Heptachlorbiphenyl (PCB Nr. 179)
2,2',3,3',5,6,6'-Heptachlorbiphenyl (PCB Nr. 179)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 52663-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR