2,3,3',4,4',5,5'-Heptachlorbiphenyl (PCB Nr. 189)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-189-C
PROOCIL-PCB-189-CEA 704 EUR
PROOCIL-PCB-189-C
2,3,3',4,4',5,5'-Heptachlorbiphenyl (PCB Nr. 189)
2,3,3',4,4',5,5'-Heptachlorbiphenyl (PCB Nr. 189)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 39635-31-9
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR