2,3,3',4,4',5',6-Heptachlorbiphenyl (PCB Nr. 191)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-191-C
PROOCIL-PCB-191-CEA 713 EUR
PROOCIL-PCB-191-C
2,3,3',4,4',5',6-Heptachlorbiphenyl (PCB Nr. 191)
2,3,3',4,4',5',6-Heptachlorbiphenyl (PCB Nr. 191)
Formel: C₁₂H₃Cl₇
Molecular Weight: 395.32 g/mol
CAS-Nummer: 74472-50-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR