2,3,3',4,5,5',6-Heptachlorbiphenyl (PCB Nr. 192)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-192-C
PROOCIL-PCB-192-CEA 713 EUR
PROOCIL-PCB-192-C
2,3,3',4,5,5',6-Heptachlorbiphenyl (PCB Nr. 192)
2,3,3',4,5,5',6-Heptachlorbiphenyl (PCB Nr. 192)
Formel: (C₆H₃Cl₂)(C₆Cl₅)
Molecular Weight: 395.32 g/mol
CAS-Nummer: 74472-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR