2,2',3,3',4,4',5,5'-Octachlorbiphenyl (PCB Nr. 194)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-194-C
PROOCIL-PCB-194-CEA 668 EUR
PROOCIL-PCB-194-C
2,2',3,3',4,4',5,5'-Octachlorbiphenyl (PCB Nr. 194)
2,2',3,3',4,4',5,5'-Octachlorbiphenyl (PCB Nr. 194)
Formel: C₁₂H₂Cl₈
Molecular Weight: 429.77 g/mol
CAS-Nummer: 35694-08-7
UN: 2315
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR