3-Chlorbiphenyl (PCB Nr. 2)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-2-C
PROOCIL-PCB-2-CEA
581
EUR
PROOCIL-PCB-2-C
3-Chlorbiphenyl (PCB Nr. 2)
3-Chlorbiphenyl (PCB Nr. 2)
Formel:
C₁₂H₉Cl Molecular Weight: 188,66 g/mol Siedepunkt: 285 °C (1013 hPa) Schmelzpunkt: 16 °C Dichte: 1,1579 g/cm³ (25 °C) |
MDL:
MFCD00013633 CAS-Nummer: 2051-61-8 EINECS: 218-126-1 UN: 2315 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...