2,3,3',4,4',5,5',6-Octachlorbiphenyl (PCB Nr. 205)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-205-C
PROOCIL-PCB-205-CEA 677 EUR
PROOCIL-PCB-205-C
2,3,3',4,4',5,5',6-Octachlorbiphenyl (PCB Nr. 205)
2,3,3',4,4',5,5',6-Octachlorbiphenyl (PCB Nr. 205)
Formel: C₁₂H₂Cl₈
Molecular Weight: 429.77 g/mol
CAS-Nummer: 74472-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR