2,2',3,3',4,4',5,5',6-Nonachlorbiphenyl (PCB Nr. 206)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-206-C
PROOCIL-PCB-206-CEA 687 EUR
PROOCIL-PCB-206-C
2,2',3,3',4,4',5,5',6-Nonachlorbiphenyl (PCB Nr. 206)
2,2',3,3',4,4',5,5',6-Nonachlorbiphenyl (PCB Nr. 206)
Formel: C₁₂HCl₉
Molecular Weight: 464.21 g/mol
CAS-Nummer: 40186-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR