2,2',4,4'-Tetrachlorbiphenyl (PCB Nr. 47)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-47-C
PROOCIL-PCB-47-CEA
677
EUR
PROOCIL-PCB-47-C
2,2',4,4'-Tetrachlorbiphenyl (PCB Nr. 47)
2,2',4,4'-Tetrachlorbiphenyl (PCB Nr. 47)
Formel:
C₁₂H₆Cl₄ Molecular Weight: 291.99 g/mol |
CAS-Nummer:
2437-79-8 EINECS: 219-444-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...