2,3-Dichlorbiphenyl (PCB Nr. 5)
Lieferant: LGC Standards PROMOCHEM
Synonyme:
PCB Nr. 5, 2,3-PCB, 1,2-Dichlor-3-phenylbenzol
CIL-PCB-5-C
PROOCIL-PCB-5-CEA
615
EUR
PROOCIL-PCB-5-C
2,3-Dichlorbiphenyl (PCB Nr. 5)
2,3-Dichlorbiphenyl (PCB Nr. 5)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,98 g/mol |
MDL:
MFCD00055244 CAS-Nummer: 16605-91-7 UN: 3432 ADR: 9,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...