2,3,3',4-Tetrachlorbiphenyl (PCB Nr. 55)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-55-C
PROOCIL-PCB-55-CEA 677 EUR
PROOCIL-PCB-55-C
2,3,3',4-Tetrachlorbiphenyl (PCB Nr. 55)
2,3,3',4-Tetrachlorbiphenyl (PCB Nr. 55)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 74338-24-2
UN: 3432
ADR: 9 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR