2,3,3',6-Tetrachlorbiphenyl (PCB Nr. 59)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-59-C
PROOCIL-PCB-59-CEA
809
EUR
PROOCIL-PCB-59-C
2,3,3',6-Tetrachlorbiphenyl (PCB Nr. 59)
2,3,3',6-Tetrachlorbiphenyl (PCB Nr. 59)
Formel:
C₁₂H₆Cl₄ Molecular Weight: 291.99 g/mol |
CAS-Nummer:
74472-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...