2,3,4,4'-Tetrachlorbiphenyl (PCB Nr. 60)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-60-C
PROOCIL-PCB-60-CEA 625 EUR
PROOCIL-PCB-60-C
2,3,4,4'-Tetrachlorbiphenyl (PCB Nr. 60)
2,3,4,4'-Tetrachlorbiphenyl (PCB Nr. 60)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 33025-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR