2,3',4,5-Tetrachlorbiphenyl (PCB Nr. 67)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-67-C
PROOCIL-PCB-67-CEA 713 EUR
PROOCIL-PCB-67-C
2,3',4,5-Tetrachlorbiphenyl (PCB Nr. 67)
2,3',4,5-Tetrachlorbiphenyl (PCB Nr. 67)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 73575-53-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR