2,3',4,5'-Tetrachlorbiphenyl (PCB Nr. 68)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-68-C
PROOCIL-PCB-68-CEA 677 EUR
PROOCIL-PCB-68-C
2,3',4,5'-Tetrachlorbiphenyl (PCB Nr. 68)
2,3',4,5'-Tetrachlorbiphenyl (PCB Nr. 68)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 73575-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR