2,3',4,6-Tetrachlorbiphenyl (PCB Nr. 69)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-69-C
PROOCIL-PCB-69-CEA 677 EUR
PROOCIL-PCB-69-C
2,3',4,6-Tetrachlorbiphenyl (PCB Nr. 69)
2,3',4,6-Tetrachlorbiphenyl (PCB Nr. 69)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 60233-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR