2,4-Dichlorbiphenyl (PCB Nr. 7)
Lieferant: LGC Standards PROMOCHEM
CIL-PCB-7-C
PROOCIL-PCB-7-CEA
615
EUR
PROOCIL-PCB-7-C
2,4-Dichlorbiphenyl (PCB Nr. 7)
2,4-Dichlorbiphenyl (PCB Nr. 7)
Formel:
C₁₂H₈Cl₂ Molecular Weight: 223,1 g/mol |
CAS-Nummer:
33284-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...