2,3',5',6-Tetrachlorbiphenyl (PCB Nr. 73)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-73-C
PROOCIL-PCB-73-CEA 713 EUR
PROOCIL-PCB-73-C
2,3',5',6-Tetrachlorbiphenyl (PCB Nr. 73)
2,3',5',6-Tetrachlorbiphenyl (PCB Nr. 73)
Formel: C₁₂H₆Cl₄
Molecular Weight: 291.99 g/mol
CAS-Nummer: 74338-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR