2,5-Dichlorbiphenyl (PCB Nr. 9)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-9-C
PROOCIL-PCB-9-CEA 615 EUR
PROOCIL-PCB-9-C
2,5-Dichlorbiphenyl (PCB Nr. 9)
2,5-Dichlorbiphenyl (PCB Nr. 9)
Formel: C₁₂H₈Cl₂
Molecular Weight: 223,1 g/mol
Siedepunkt: 182 °C (1013 hPa)
Schmelzpunkt: 19 °C
CAS-Nummer: 34883-39-1
UN: 2315
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR