2,2',4,4',5-Pentachlorbiphenyl (PCB Nr. 99)

Lieferant: LGC Standards PROMOCHEM

CIL-PCB-99-C
PROOCIL-PCB-99-CEA 677 EUR
PROOCIL-PCB-99-C
2,2',4,4',5-Pentachlorbiphenyl (PCB Nr. 99)
2,2',4,4',5-Pentachlorbiphenyl (PCB Nr. 99)
Formel: C₁₂H₅Cl₅
Molecular Weight: 326.44 g/mol
CAS-Nummer: 38380-01-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR