Acridin-[D9] Isotope_(98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: Acridine-[²H₉]

CILDLM-849-0.1 CILDLM-849-0.5
PROOCILDLM-849-0.1EA 515 EUR
PROOCILDLM-849-0.1 PROOCILDLM-849-0.5
Acridin-[D9] Isotope_(98% D)
Acridin-[D9]
Formel: C₁₃D₉N
Molecular Weight: 188.15 g/mol
CAS-Nummer: 34749-75-2
UN: 2713
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR