Bis(2-ethylhexyl)phthalat-[Ring-D4] (98% D)

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: Bis(2-ethylhexyl) phthalate-[ring-²H₄]

CILDLM13680.25
PROOCILDLM13680.25EA 1380 EUR
PROOCILDLM13680.25
Bis(2-ethylhexyl)phthalat-[Ring-D4] (98% D)
Bis(2-ethylhexyl)phthalat-[Ring-D4]
Formel: C₂₄H₃₄D₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00143759
CAS-Nummer: 93951-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR