Benzylbutylphthalat-[Ring-D4] (98% D)
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Benzyl butyl phthalate-[ring-²H₄]
CILDLM13690.1
PROOCILDLM13690.1EA
495
EUR
PROOCILDLM13690.1
Benzylbutylphthalat-[Ring-D4] (98% D)
Benzylbutylphthalat-[Ring-D4]
Formel:
C₁₉H₁₆D₄O₄ |
CAS-Nummer:
93951-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...