Di-n-octylphthalat-[Ring-D4] (98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: Di-n-octyl phthalate-[ring-²H₄]

CILDLM16300.1
PROOCILDLM16300.1EA 534 EUR
PROOCILDLM16300.1
Di-n-octylphthalat-[Ring-D4] (98% D)
Di-n-octylphthalat-[Ring-D4]
Formel: C₂₄H₃₄D₄O₄
MDL: MFCD00144150
CAS-Nummer: 93952-13-7
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR