Benzo[j]fluoranthen

Lieferant: LGC Standards PROMOCHEM

CILULM241125
PROOCILULM241125EA 672 EUR
PROOCILULM241125
Benzo[j]fluoranthen
Benzo[j]fluoranthen
Formel: C₂₀H₁₂
CAS-Nummer: 205-82-3
EINECS: 205-910-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR