Benzo[b]fluoranthen
Lieferant: LGC Standards PROMOCHEM
CILULM24160.1
PROOCILULM24160.1EA
292
EUR
PROOCILULM24160.1
Benzo[b]fluoranthen
Benzo[b]fluoranthen
Formel:
C₂₀H₁₂ |
MDL:
MFCD00010582 CAS-Nummer: 205-99-2 EINECS: 205-911-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...