Dibenzo[a,j]acridin

Lieferant: LGC Standards PROMOCHEM

CILULM388425
PROOCILULM388425EA 728 EUR
PROOCILULM388425
Dibenzo[a,j]acridin
Dibenzo[a,j]acridin
Formel: C₂₁H₁₃N
Molecular Weight: 279.34 g/mol
CAS-Nummer: 224-42-0
UN: 1294
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR