Di-n-octylphthalat
Lieferant: LGC Standards PROMOCHEM
CILULM61291.2
PROOCILULM61291.2EA
190
EUR
PROOCILULM61291.2
Di-n-octylphthalat
Di-n-octylphthalat
Formel:
C₂₄H₃₈O₄ |
MDL:
MFCD00015292 CAS-Nummer: 117-84-0 EINECS: 204-214-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...