Benzyldimethyldodecylammoniumbromid

Lieferant: LGC Standards PROMOCHEM

Synonyme: Benzyldodecyldimethylammonium bromide, Benzalkonium bromide, N-Benzyl-N,N-dimethyldodecan-1-aminium bromide

FPC0141
PROOFPC0141EA 206 EUR
PROOFPC0141
Benzyldimethyldodecylammoniumbromid
Benzyldimethyldodecylammoniumbromid
Formel: C₂₁H₃₈BrN
Molecular Weight: 384,44 g/mol
MDL: MFCD00011768
CAS-Nummer: 7281-04-1
EINECS: 230-698-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR