Viscosity test standard for PTP, VHG
Lieferant: LGC Standards PROMOCHEM
G-VPTPVISC-50
PROOG-VPTPVISC-50EA 82 EUR
PROOG-VPTPVISC-50
Viscosity test standard for PTP, VHG
Standards Viskosimetrie-Standards
This proficiency testing program (PTP) was specially designed to give analytical laboratories instant validation while measuring viscosity in new and used oils.
Standards: Traceable to CRM. ISO/IEC 17043 accredited.