Tetrachlorbrenzcatechin
Lieferant: LGC Standards PROMOCHEM
Synonyme:
Tetrachlorbenzol-1,2-diol
HXC004
PROOHXC004EA
289
EUR
PROOHXC004
Tetrachlorbrenzcatechin
Tetrachlorbrenzcatechin
Formel:
C₆H₂Cl₄O₂ Molecular Weight: 247,89 g/mol |
MDL:
MFCD00002190 CAS-Nummer: 1198-55-6 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...