Ampicillin Natriumsalz
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
ICRS0212
PROOICRS0212EA
226
EUR
PROOICRS0212
Ampicillin Natriumsalz
Ampicillin Natriumsalz
Formel:
C₁₆H₁₈N₃NaO₄S Molecular Weight: 371.39 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00064313 CAS-Nummer: 69-52-3 EINECS: 200-708-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...