Ampicillin Trihydrat
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
ICRS0213
PROOICRS0213EA
226
EUR
PROOICRS0213
Ampicillin Trihydrat
Ampicillin Trihydrat
Formel:
C₁₆H₁₉N₃O₄S·3H₂O Lagertemperatur: Kühlschrank |
MDL:
MFCD00072036 CAS-Nummer: 7177-48-2 EINECS: 200-709-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...