Chlortetracyclin Hydrochlorid
Lieferant: LGC Standards PROMOCHEM
|
Gefahr
|
Synonyme:
Chlortetracycline HCI, Aureomycin, 7-Chlorotetracycline hydrochloride, (4S,4aS,5aS,6S,12aS)-7-Chloro-4-(dimethylamino)-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrochloride
ICRS0249
PROOICRS0249EA
226
EUR
PROOICRS0249
Chlortetracyclin Hydrochlorid
Chlortetracyclin Hydrochlorid
Formel:
C₂₂H₂₃ClN₂O₈·HCl Molecular Weight: 515.34 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD26142699 CAS-Nummer: 64-72-2 EINECS: 200-591-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...