Cisplatin

Lieferant: LGC Standards PROMOCHEM

Synonyme: cis-Diammindichloridoplatin(II), Platinol

ICRS0258
PROOICRS0258EA 226 EUR
PROOICRS0258
Cisplatin
cis-Platin
Formel: H₆Cl₂N₂Pt
Molecular Weight: 300,05 g/mol
Dichte: 3,738 g/cm³ (20 °C)
MDL: MFCD00011623
CAS-Nummer: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR