Dapson
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
ICRS0265
PROOICRS0265EA
226
EUR
PROOICRS0265
Dapson
Dapson
Formel:
C₁₂H₁₂N₂O₂S |
MDL:
MFCD00007887 CAS-Nummer: 80-08-0 EINECS: 201-248-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...