Digoxin
Lieferant: LGC Standards PROMOCHEM
ICRS0289
PROOICRS0289EA
226
EUR
PROOICRS0289
Digoxin
Digoxin
Formel:
C₄₁H₆₄O₁₄ Molecular Weight: 780.95 g/mol |
MDL:
MFCD00003674 CAS-Nummer: 20830-75-5 EINECS: 244-068-1 UN: 2811 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...