5-Fluorcytosin

Lieferant: LGC Standards PROMOCHEM

Synonyme: Flucytosin, 4-Amino-5-fluor-2-hydroxypyrimidin, 4-Amino-5-fluor-2-pyrimidinol, 5-FC

ICRS0310
PROOICRS0310EA 227 EUR
PROOICRS0310
5-Fluorcytosin
5-Fluorcytosin
Formel: C₄H₄FN₃O
Molecular Weight: 129,09 g/mol
Siedepunkt: 298 °C (1013 hPa)
Schmelzpunkt: 165 °C
MDL: MFCD00006035
CAS-Nummer: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR