3-(3,4-Dihydroxyphenyl)-L-alanin

Lieferant: LGC Standards PROMOCHEM

Synonyme: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

ICRS0334
PROOICRS0334EA 226 EUR
PROOICRS0334
3-(3,4-Dihydroxyphenyl)-L-alanin
3-(3,4-Dihydroxyphenyl)-L-alanin
Formel: C₉H₁₁NO₄
Molecular Weight: 197,19 g/mol
MDL: MFCD00002598
CAS-Nummer: 59-92-7
EINECS: 200-445-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR