Methotrexat

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: (S)-2-(4-(((2,4-Diaminopteridin-6-yl)methyl)(methyl)amino)benzamido)pentanedioic acid

ICRS0345
PROOICRS0345EA 226 EUR
PROOICRS0345
Methotrexat
Methotrexat
Formel: C₂₀H₂₂N₈O₅
Molecular Weight: 454,44 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00150847
CAS-Nummer: 59-05-2
EINECS: 200-413-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR