Nystatin

Lieferant: LGC Standards PROMOCHEM
Achtung

ICRS0369
PROOICRS0369EA 226 EUR
PROOICRS0369
Nystatin
Nystatin
Formel: C₄₇H₇₅NO₁₇
Lagertemperatur: Gefrierschrank
MDL: MFCD25541651
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR