Paracetamol
Lieferant: LGC Standards PROMOCHEM
Synonyme:
4'-Hydroxyacetanilid, 4-Acetamidophenol, Acetaminophen, N-(4-Hydroxyphenyl)acetamid, Paracetamolum, APAP, Ethansäure(4-hydroxyphenyl)amid, N-Acetyl-4-aminophenol
ICRS0377
PROOICRS0377EA
226
EUR
PROOICRS0377
Paracetamol
Paracetamol
Formel:
C₈H₉NO₂ Molecular Weight: 151,16 g/mol Siedepunkt: 388 °C (1013 hPa) Schmelzpunkt: 168 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002328 CAS-Nummer: 103-90-2 EINECS: 203-157-5 Merck Index: 13,00048 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...