5,5-Diphenylhydantoin
Lieferant: LGC Standards PROMOCHEM
ICRS0387
PROOICRS0387EA
226
EUR
PROOICRS0387
5,5-Diphenylhydantoin
5,5-Diphenylhydantoin
Formel:
C₁₅H₁₂N₂O₂ |
MDL:
MFCD00005264 CAS-Nummer: 57-41-0 EINECS: 200-328-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...