Procain Hydrochlorid

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2-Diethylaminoethyl 4-aminobenzoate hydrochloride

ICRS0398
PROOICRS0398EA 226 EUR
PROOICRS0398
Procain Hydrochlorid
Procain Hydrochlorid
Formel: C₁₃H₂₁ClN₂O₂
MDL: MFCD00013000
CAS-Nummer: 51-05-8
EINECS: 200-077-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR