6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon
Lieferant: LGC Standards PROMOCHEM
|
Achtung
|
Synonyme:
Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidin, 4-Propyl-2-thiouracil
ICRS0403
PROOICRS0403EA
226
EUR
PROOICRS0403
6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon
6-Propyl-2-thiouracil
Formel:
C₇H₁₀N₂OS Molecular Weight: 170,24 g/mol Siedepunkt: 448 °C (1013 hPa) Schmelzpunkt: 215…218 °C |
MDL:
MFCD00006041 CAS-Nummer: 51-52-5 EINECS: 200-103-2 Merck Index: 12,08054 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...