Saccharin
Lieferant: LGC Standards PROMOCHEM
Synonyme:
o-Sulphobenzimide, o-Benzoic sulphimide
ICRS0411
PROOICRS0411EA
223
EUR
PROOICRS0411
Saccharin
Saccharin
Formel:
C₇H₅NO₃S Molecular Weight: 183,19 g/mol |
MDL:
MFCD00005866 CAS-Nummer: 81-07-2 EINECS: 201-321-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...